Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8984

Sigma-Aldrich

Monoclonal Anti-Protein Tyrosine Phosphatase μ antibody produced in mouse

clone SBK10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PTPμ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SBK10, monoclonal

form

buffered aqueous solution

mol wt

antigen 200 kDa

species reactivity

rat, mouse, human, bovine

technique(s)

immunoprecipitation (IP): suitable
western blot (chemiluminescent): 1-10 μg/mL

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRM(5797)
mouse ... Ptprm(19274)
rat ... Ptprm(29616)

General description

Among the post-translational modifications, phosphorylation is a vital regulatory mechanism of key proteins involved in specific pathways. Reverse phosphorylation has become recognized as the key process of regulation of gene expression, cellular proliferation, differentiation in Eukaryotes. The protein phosphatases can be divided into two main groups: protein tyrosine phosphatases (PTPs) and protein serine/threonine phosphatases (PPs) which remove phosphate from proteins/peptides containing phosphotyrosine (pTyr) or phosphoserine/phosphothreonine (pSer/pThr), respectively. Several of the PTPs are known to control the function of growth factor receptors, many of which are tyrosine kinases encoded by oncogenes. PTPmu participates in homophilic binding of extracellular surface of adjacent cells. PTPmu is reported to be downregulated in glioblastoma in which it regulates cell migration and growth factor-independent survival
Monoclonal Anti-Protein Tyrosine Phosphatase mu recognizes PTPmu isoforms in all mammalian species.

Immunogen

peptide corresponding to amino acid residues 42-60 of PTPμ.

Application

Anti-protein tyrosine phosphatase mu may be used for detection by immunoblotting at a working concentration of 1 to 10 μg/mL. The antibody is suitable for immunoprecipitation.

Physical form

Solution in phosphate buffered saline with 0.08% sodium azide.

Preparation Note

Purified from tissue culture supernatant using Protein G.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Gjörloff-Wingren et al.
European journal of immunology, 30(8), 2412-2421 (2000-08-15)
A high protein tyrosine phosphatase (PTPase) activity is required to maintain circulating T lymphocytes in a resting phenotype, and to limit the initiation of T cell activation. We report that 15 of the currently known 24 intracellular PTPases are expressed
Adam M Burgoyne et al.
Cancer research, 69(17), 6960-6968 (2009-08-20)
Glioblastoma multiforme (GBM), the most common malignant primary brain tumor, represents a significant disease burden. GBM tumor cells disperse extensively throughout the brain parenchyma, and the need for tumor-specific drug targets and pharmacologic agents to inhibit cell migration and dispersal
S M Brady-Kalnay et al.
The Journal of biological chemistry, 269(45), 28472-28477 (1994-11-11)
The receptor-type protein tyrosine phosphatase PTP mu comprises an extracellular segment containing a MAM domain, an immunoglobulin domain and four fibronectin type III repeats, a transmembrane segment, and two intracellular PTP domains. We have previously shown that PTP mu binds
Adam M Burgoyne et al.
Neuro-oncology, 11(6), 767-778 (2009-03-24)
The cell-surface receptor protein tyrosine phosphatase mu (PTPmu) is a homophilic cell adhesion molecule expressed in CNS neurons and glia. Glioblastomas (GBMs) are the highest grade of primary brain tumors with astrocytic similarity and are characterized by marked dispersal of
Ronglin Wang et al.
Cell death & disease, 9(6), 693-693 (2018-06-09)
MiRNAs, a group of powerful modulator of gene expression, participate in multiple cellular processes under physiological and pathological conditions. Emerging evidence shows that Drosha, which controls the initial step in canonical miRNA biogenesis, is involved in modulating cell survival and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service