Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P8977

Sigma-Aldrich

AutoPAGE Plus 4.5% High Throughput Acrylamide Premix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

storage temp.

2-8°C

Quality Level

application

AutoPAGE Plus is a specially formulated sequencing gel solution that provides read lengths of 700–800 bases in as little as seven hours, allowing you to run up to three gels in a 24 hour period (Figure 1). When maximum throughput is not your concern, AutoPAGE Plus can also be used for 10 to 12 hour 2X runs providing read lengths of 900 base pairs or more. In addition to the long reads, the superior resolution of AutoPAGE Plus generates higher quality sequencing data. PHRED scores generated in a blind analysis of three commercially available solutions show the dramatic difference in high quality and very high quality data (Figure 2).

While providing the most reliable and reproducible data, AutoPAGE Plus can increase your throughput by up to 50% or reduce the time you have to wait to analyze your results.

Features and Benefits

  • 700-800 bases in as little as seven hours
  • Also compatible with 2× overnight runs yielding 900-1000 bases
  • Superior resolution
  • Compatible with dye primer and dye terminator chemistry including BigDye™ chemistry
  • Performance tested with ABI 377 instruments
  • No decrease in read length or resolution when used on instruments with 96 lane upgrade
  • No filtering or degassing necessary

Physical form

Stable for one year at 2-8 °C

Legal Information

AutoPAGE is a trademark of Sigma-Aldrich Co. LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service