Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8912

Sigma-Aldrich

R-Phycoerythrin from Porphyra tenera “Nori”

lyophilized powder

Synonym(s):

PE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

biological source

algae (Porphyra tenera)

form

lyophilized powder

mol wt

~255 kDa

composition

Protein, ~30% Lowry

concentration

~30% protein (Lowry)

color

pink to purple

fluorescence

λex 488 nm; λem 578 nm(lit.)

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

For labeling proteins to make fluorescent probes
R-phycoerythrin from Porphyra tenera has been used in ORAC assays to measure the antioxidant capacity of various nutrients.

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing sucrose, dithioerythritol and sodium azide

Pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperoxia-induced changes in antioxidant capacity and the effect of dietary antioxidants.
Cao, G.
Journal of Applied Physics, 86, 1817-1822 (1999)
The use of phycobiliproteins as fluorescent labels in immunoassay.
M N Kronick
Journal of immunological methods, 92(1), 1-13 (1986-08-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service