Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8872

Sigma-Aldrich

Anti-Pituitary Adenylate Cyclase Activating Polypeptide Type 1 Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PACAP Receptor Type 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 7 μg/mL using human pituitary

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ADCYAP1R1(117)

Immunogen

synthetic peptide corresponding to the N-terminal extracellular domain of human pituitary adenylate cyclase activating polypeptide type 1 receptor (PACAP), conjugated to KLH. The immunizing peptide has 94% homology with the rat and mouse gene.

Target description

Pituitary Adenylate Cyclase-Activating Polypeptide is present not only in the central nervous system but also in peripheral tissues,including the gastrointestinal tract, adrenal gland, andtestes. Its actions include the stimulation of secretion ofgr

Physical form

Solution in phosphate buffered saline containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ildiko Somogyi et al.
Annals of the New York Academy of Sciences, 1163, 521-523 (2009-05-22)
By means of radioimmunoassay, we studied the concentration of pituitary adenylate cyclase-activating polypeptide (PACAP)-like proteins in intact and regenerating earthworms. Transection of animals increased the concentration of PACAP-like compounds in coelomocytes, and a decreasing rostrocaudal gradient was detected in the
P Egri et al.
Endocrinology, 157(6), 2356-2366 (2016-04-06)
The hypothalamic activation of thyroid hormones by type 2 deiodinase (D2), catalyzing the conversion of thyroxine to T3, is critical for the proper function of the hypothalamo-pituitary-thyroid (HPT) axis. Regulation of D2 expression in tanycytes alters the activity of the
Jacqueline Hill et al.
American journal of physiology. Cell physiology, 303(3), C257-C266 (2012-05-18)
Neuroendocrine adrenal medullary chromaffin cells receive synaptic excitation through the sympathetic splanchnic nerve to elicit catecholamine release into the circulation. Under basal sympathetic tone, splanchnic-released acetylcholine evokes chromaffin cells to fire action potentials, leading to synchronous phasic catecholamine release. Under
Tamás Juhász et al.
International journal of molecular sciences, 16(8), 17344-17367 (2015-08-01)
Pituitary adenylate cyclase activating polypeptide (PACAP) is a neurohormone exerting protective function during various stress conditions either in mature or developing tissues. Previously we proved the presence of PACAP signaling elements in chicken limb bud-derived chondrogenic cells in micromass cell
Nóra Krajcs et al.
Journal of molecular neuroscience : MN, 57(4), 492-500 (2015-07-04)
In this study, we investigate the potentiating effect of PACAP27 on cholinergic neuromuscular transmission in the recently discovered flexor muscles of the land snail, Helix pomatia. Using immunohistochemistry, we show that PACAP and PAC1 receptors are present in nerve fibers

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service