Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8871

Sigma-Aldrich

Anti-Phosphodiesterase 6β antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PDE6β

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

bovine, mouse

technique(s)

western blot: 2 μg/mL using mouse retinal extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

mouse ... Pde6b(18587)

Immunogen

synthetic peptide which corresponds to amino acid residues 20-36 from mouse and bovine PDE6β protein.

Physical form

Solution in phosphate buffered saline containing 1 mg/mL BSA and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service