About This Item
assay
≥95%
form
powder
color
yellow
storage temp.
−20°C
SMILES string
CC1C(OC(=O)\C=C(C)\C=C\C=C(C)\C=C\C2=C(C)CCCC2(C)C)C3(OC(C)=O)C(C4C=C(CO)CC5(O)C(C=C(C)C5=O)C14O)C3(C)C
InChI
1S/C42H56O8/c1-24(16-17-31-26(3)15-12-18-38(31,7)8)13-11-14-25(2)19-34(45)49-37-28(5)41(48)32(35-39(9,10)42(35,37)50-29(6)44)21-30(23-43)22-40(47)33(41)20-27(4)36(40)46/h11,13-14,16-17,19-21,28,32-33,35,37,43,47-48H,12,15,18,22-23H2,1-10H3/b14-11+,17-16+,24-13+,25-19+/t28-,32+,33-,35-,37-,40-,41-,42-/m1/s1
InChI key
GTNCUWBKHZZECN-IXTPSRJDSA-N
Biochem/physiol Actions
signalword
Danger
Hazard Classifications
Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service