Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8746

Sigma-Aldrich

Anti-Phosphodiesterase 6δ antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PDE6δ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 17 kDa

species reactivity

bovine, human, sheep

technique(s)

immunohistochemistry: 1 μg/mL using bovine retinal sections
western blot: 2 μg/mL using bovine retinal extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PDE6D(5147)

Specificity

Does not detect other PDE6 isoforms.

Immunogen

synthetic peptide corresponding to amino acid residues 1-16 from mouse and human PDE6δ protein.

Physical form

Solution in phosphate buffered saline containing 1 mg/mL BSA and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service