Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

P8732

Sigma-Aldrich

Anti-Potassium Channel Kv1.2 (Dendrotoxin-Sensitive Delayed Rectifier; BK2; RCK5; NGK1; RK2; RAK) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, mouse

technique(s)

western blot (chemiluminescent): 1:200

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Kcna2(16490)
rat ... Kcna2(25468)

Specificity

Recognizes a full-length Kv1.2 protein. Does not cross-react with other potassium channel antigens.

Immunogen

GST fusion protein of a C-terminal part of rat potassium channel Kv1.2 corresponding to amino acids 417-498.

Physical form

Lyophilized at ~0.2 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service