Key Documents
P8687
Finnpipette® PDP tips
maximum volume 200 μL
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
200 μL
sterility:
non-sterile
packaging:
pack of 960 ea (Hinged rack in box)
manufacturer/tradename:
Finn 9403000
material
blue
sterility
non-sterile
packaging
pack of 960 ea (Hinged rack in box)
manufacturer/tradename
Finn 9403000
L
4.9 cm
maximum volume
200 μL
volume
200 μL
color
blue
General description
Polypropylene barrels and polyethylene pistons. Idael for pipetting viscous liquid. Designed for Finnpipette PDP
Packaging
10 boxes of 96 tips/ea. in a autoclavable rack with hinged top.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)