Key Documents
P8552
Sigma-Aldrich
Pipette canisters
For 1 - 10 mL pipettes
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
size
67 mm × 67 mm × 406 mm
General description
Heavy gauge, satin finish stainless steel. Square cross-section permits easy autoclaving and storage. Silicone pads inside top and bottom minimize chipping of pipette tips.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Customers Also Viewed
Slide 1 of 1