Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8363

Sigma-Aldrich

Poly(A)+ RNA human

from human(normal adult)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.51

biological source

human (normal adult)

Quality Level

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Packaging

Each vial contains 5 μg of poly(A)+ RNA in DEPC treated water

Other Notes

Human mRNA is isolated from total RNA by affinity chromatography using oligo(dT) columns. Following isolation, the integrity of the mRNA is verified on a denaturing agarose gel. All mRNAs show very high efficiencies in reverse transcription.

Legal Information

Manufactured by BioChain, Inc.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service