Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

P7982

Sigma-Aldrich

Anti-P2X2 Purinergic Receptor antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot (chemiluminescent): 1:200

UniProt accession no.

storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide corresponding to amino acids 457-472 of rat P2X2 (Swiss-Prot P49653) (with additional N-terminal cysteine).

Physical form

Lyophilized at ~0.3 mg/ml from a solution of phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makoto Ohmoto et al.
eNeuro, 10(4) (2023-04-13)
Taste cells are maintained by continuous turnover throughout a lifetime, yet the mechanisms of taste cell differentiation, and how taste sensations remain constant despite this continuous turnover, remain poorly understood. Here, we report that a transcription factor Etv1 (also known
Kengo Nomura et al.
Neuron, 106(5), 816-829 (2020-04-02)
Sodium taste regulates salt intake. The amiloride-sensitive epithelial sodium channel (ENaC) is the Na+ sensor in taste cells mediating attraction to sodium salts. However, cells and intracellular signaling underlying sodium taste in taste buds remain long-standing enigmas. Here, we show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service