Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7806

Supelco

Sigma Pseudo Narcotic Scent Heroin formulation

solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107

form

solution

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Substitute for controlled substances in the training of narcotics detector dogs. Mimics the odor of heroin.

Legal Information

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service