P7619
Prepro-Atrial Natriuretic Peptide Fragment 26-55 human
≥95% (HPLC)
Synonym(s):
Prepro-hANF fragment 26-55
About This Item
Recommended Products
Product Name
Prepro-Atrial Natriuretic Peptide Fragment 26-55 human, ≥95% (HPLC)
Quality Level
assay
≥95% (HPLC)
form
powder
color
white
UniProt accession no.
storage temp.
−20°C
SMILES string
CSCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CO)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](CCSC)NC(=O)[C@@H]2CCCN2C(=O)[C@@H](N)CC(N)=O)C(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc4c[nH]cn4)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCSC)C(=O)N5CCC[C@H]5C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(O)=O)C(O)=O
InChI
1S/C152H236N38O51S3/c1-72(2)53-93(134(222)167-88(38-41-116(199)200)128(216)166-87(37-40-115(197)198)127(215)164-85(29-21-23-46-153)125(213)171-92(45-52-244-17)151(239)190-49-26-32-110(190)148(236)185-97(57-76(9)10)136(224)168-89(39-42-117(201)202)129(217)186-107(152(240)241)68-121(209)210)174-141(229)100(60-82-69-160-71-161-82)179-145(233)106(67-120(207)208)184-138(226)96(56-75(7)8)173-137(225)95(55-74(5)6)175-142(230)103(64-114(159)196)180-126(214)86(30-22-24-47-154)165-139(227)98(58-80-27-19-18-20-28-80)178-144(232)105(66-119(205)206)183-130(218)90(43-50-242-15)169-135(223)94(54-73(3)4)176-143(231)104(65-118(203)204)172-123(211)78(13)162-133(221)102(63-113(158)195)182-146(234)108(70-191)187-149(237)122(77(11)12)188-124(212)79(14)163-132(220)101(62-112(157)194)181-140(228)99(59-81-33-35-83(192)36-34-81)177-131(219)91(44-51-243-16)170-147(235)109-31-25-48-189(109)150(238)84(155)61-111(156)193/h18-20,27-28,33-36,69,71-79,84-110,122,191-192H,21-26,29-32,37-68,70,153-155H2,1-17H3,(H2,156,193)(H2,157,194)(H2,158,195)(H2,159,196)(H,160,161)(H,162,221)(H,163,220)(H,164,215)(H,165,227)(H,166,216)(H,167,222)(H,168,224)(H,169,223)(H,170,235)(H,171,213)(H,172,211)(H,173,225)(H,174,229)(H,175,230)(H,176,231)(H,177,219)(H,178,232)(H,179,233)(H,180,214)(H,181,228)(H,182,234)(H,183,218)(H,184,226)(H,185,236)(H,186,217)(H,187,237)(H,188,212)(H,197,198)(H,199,200)(H,201,202)(H,203,204)(H,205,206)(H,207,208)(H,209,210)(H,240,241)/t78-,79-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,122-/m0/s1
InChI key
PCSAOHPOZFJINS-BGLQADKRSA-N
Gene Information
human ... NPPA(4878)
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service