Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7587

Sigma-Aldrich

Phenyl isothiocyanate solution

5% in heptane

Synonym(s):

PITC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NCS
CAS Number:
Molecular Weight:
135.19
Beilstein/REAXYS Number:
471392
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

concentration

5% in heptane

storage temp.

2-8°C

SMILES string

S=C=Nc1ccccc1

InChI

1S/C7H5NS/c9-6-8-7-4-2-1-3-5-7/h1-5H

InChI key

QKFJKGMPGYROCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

24.8 °F

flash_point_c

-4 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Gheshlaghi et al.
Analytical biochemistry, 383(1), 93-102 (2008-08-30)
Modified resolution and overall separation factors used to quantify the separation of complex chromatography systems are described. These factors were proven to be applicable to the optimization of amino acid resolution in reverse-phase (RP) HPLC chromatograms. To optimize precolumn derivatization
Misako Aito-Inoue et al.
Journal of agricultural and food chemistry, 54(15), 5261-5266 (2006-07-20)
For the isolation and detection of food-derived peptides in blood, an approach based on the derivatization of peptides with phenyl isothiocyanate (PITC) was developed. This approach allows hydrophilic peptides to be resolved and specifically detected by reversed-phase (RP) HPLC. For
Fantao Meng et al.
Bioconjugate chemistry, 19(7), 1352-1360 (2008-06-25)
Successful and cost-effective PEGylation protocols require pure functionalized PEG reagents, which can be synthesized by simple and efficient procedures, exhibit high stability against hydrolysis, and maintain a level of reactivity with protein functional groups under mild reaction conditions. PEG-phenyl-isothiocyanate (PIT-PEG)
R Thippeswamy et al.
Journal of agricultural and food chemistry, 54(19), 7014-7019 (2006-09-14)
Two liquid chromatographic methods that involve precolumn derivatization with o-phthaladehyde (OPA) and phenylisothiocyanate (PITC) with fluorescence and diode array UV detection for the determination of theanine have been developed. The chromatographic separations were achieved by reverse-phase high-performance liquid chromatography using
Tomoyuki Oe et al.
Rapid communications in mass spectrometry : RCM, 24(2), 173-179 (2009-12-17)
This manuscript describes the results of a preliminary experiment performed as 'proof of concept' of a novel approach to absolute quantitation of proteins without the use of standard proteins. Absolute quantitation remains a challenging issue in the proteomics field. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service