Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7542

Sigma-Aldrich

Carestream® Processing chemicals

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
45141500

Reconstitution

Two-part liquid concentrate to make the indicated volume.

Other Notes

Can also be used as a replenisher to extend the life of the original solution.

Legal Information

Kodak is a registered trademark of Eastman Kodak Company

Pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service