Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7520

Sigma-Aldrich

Tree/shrub pollens from Populus nigra italica (Lombardy poplar)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solid

Storage temp.

2-8°C

Pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service