The conjugates will identify and quantitate cells containing intracellular forms of human IL-4.
Immunogen
recombinant human IL-4
Biochem/physiol Actions
Interleukin-4 (IL-4) is a T cell derived chemokine that stimulates the Th2 mediated immune response and proliferation of B cells. The effects of IL-4 are mediated by two types of receptors, type I receptor consisting of IL-4Rα and common γ chain and type II receptor composed of IL-4Rα and IL-13Rα1. The signaling stimulated by IL-4 leads to activation of JAK/STAT6 and IRS-mediated PI3K/Akt pathway. IL-4 is responsible for endocytic activity of macrophages, chemotaxis of leukocytes in response to inflammation, angiogenesis and regulation of nitric oxide metabolism in macrophages. Conjugation of R-Phycoerythrin to the antibody enables its use in immunofluorescent techniques.
Physical form
25 μg of antibody in 1.0 mL of saline containing up to 0.5% bovine serum albumin. Contains 0.1% sodium azide as a preservative.
Preparation Note
Purified antibody is conjugated to R-phycoerythrin (PE).
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Although macrophages were originally recognized as major immune effector cells, it is now appreciated that they also play many important roles in the maintenance of tissue homeostasis, and are involved in a variety of pathological conditions including cancer. Several studies
The Journal of allergy and clinical immunology, 105(6 Pt 1), 1063-1070 (2000-06-16)
In the past several years, extensive studies on the mechanisms underlying IL-4 and IL-13 signaling have enabled us to gain insight into how these cytokines regulate immune responses. Because both IL-4 and IL-13 use the IL-4Ralpha as a receptor component
The mechanism of differentiation of naïve T cells to a variety of effector lineages, but particularly to T helper type 1 (Th1) and Th2 cells, has been the subject of intense scrutiny over the past two decades. Studies have revealed
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.