Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6982

Sigma-Aldrich

Prolactin-releasing Peptide human

≥97% (HPLC)

Synonym(s):

PrRP31

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C160H252N56O42S
Molecular Weight:
3664.13
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

assay

≥97% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PRLH(51052)

Amino Acid Sequence

Ser-Arg-Thr-His-Arg-His-Ser-Met-Glu-Ile-Arg-Thr-Pro-Asp-Ile-Asn-Pro-Ala-Trp-Tyr-Ala-Ser-Arg-Gly-Ile-Arg-Pro-Val-Gly-Arg-Phe-NH2

Biochem/physiol Actions

Endogenous agonist for the hGR3 receptor.
Endogenous agonist for the hGR3 receptor. Releases prolactin from pituitary cultures and LH-RH from hypothalamic explants.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service