P6893
Poly-L-lysine–Agarose
mol wt 4,000-15,000, saline suspension
About This Item
Recommended Products
form
saline suspension
mol wt
4,000-15,000
extent of labeling
1-2 mg per mL
technique(s)
protein purification: suitable
color
white to off-white
matrix
cross-linked 4% beaded agarose
matrix activation
cyanogen bromide
matrix attachment
ε-amino
matrix spacer
1 atom
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Physical form
Disclaimer

Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service