Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6665

Sigma-Aldrich

Poly(Pro-Gly-Pro)

mol wt 2,000-10,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.26

mol wt

2,000-10,000

anion traces

bromide (Br-): ≤5%

application(s)

cell analysis

storage temp.

−20°C

Analysis Note

Molecular weight based on MALLS.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C K Mukhopadhyay et al.
The Journal of biological chemistry, 269(48), 30200-30205 (1994-12-02)
In this paper we demonstrate that in the absence of free metal ions, active oxygen species, generated by activated macrophages or xanthine/xanthine oxidase (XOD), carry out oxidative degradation of collagen fibrils type I in conjunction with proteases. The collagen degradation
M G Paterlini et al.
Biopolymers, 35(6), 607-619 (1995-06-01)
The sequence-dependent local destabilization in the interior of the collagen triple helix has been evaluated by means of conformational energy computations. Using a model poly(Gly-Pro-Pro) triple helix as the reference state, a method was developed for generating local loops, i.e.
M F Hoylaerts et al.
The Biochemical journal, 306 ( Pt 2), 453-463 (1995-03-01)
In the absence of high shear forces, the in vitro binding of human von Willebrand factor (vWF) to its platelet receptor glycoprotein Ib (GPIb) can be promoted by two well-characterized mediators, botrocetin and ristocetin. Using purified vWF and GPIb, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service