Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6622

Sigma-Aldrich

Anti-Purinergic Receptor P2Y6 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunocytochemistry: 3 μg/mL using human placenta, villi
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 3 μg/mL using human placenta, villi

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... P2RY6(5031)
mouse ... P2ry6(233571)
rat ... P2ry6(117264)

Immunogen

synthetic peptide corresponding to the C-terminal domain of human P2Y6 purinergic receptor, conjugated to KLH. The immunizing peptide has 100% homology to the rat and mouse gene.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service