P6615
17-Phenyl-tri-norprostaglandin D2
≥98%, methyl acetate solution
About This Item
Quality Level
assay
≥98%
form
methyl acetate solution
storage temp.
−20°C
SMILES string
O[C@@H](CCc1ccccc1)\C=C\[C@@H]2[C@@H](C\C=C/CCCC(O)=O)[C@@H](O)CC2=O
InChI
1S/C23H30O5/c24-18(13-12-17-8-4-3-5-9-17)14-15-20-19(21(25)16-22(20)26)10-6-1-2-7-11-23(27)28/h1,3-6,8-9,14-15,18-21,24-25H,2,7,10-13,16H2,(H,27,28)/b6-1-,15-14+/t18-,19+,20+,21-/m0/s1
InChI key
OAQGPAZDRCBBLD-YTCWWFNZSA-N
Application
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
8.6 °F - closed cup
flash_point_c
-13 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service