Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P6368

Sigma-Aldrich

Anti-phospho-Paxillin (pTyr31) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 68 kDa

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: 1:1000 using NMμMG cells treated with TGF-β or chick embryo fibroblasts

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PXN(5829)

Specificity

Specifically recognizes endogenous and recombinant forms of human wild type paxillin phosphorylated on tyrosine 31. It does not cross react with recombinant Y31F mutant proteins.

Immunogen

synthetic phosphorylated peptide derived from the region of paxillin that contains a phosphate on tyrosine 31. This region is conserved between human and chick.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 with 50% glycerol, 1.0 mg/ml BSA (IgG and protease free) and 0.05% sodium azide.

Analysis Note

The amount of the reagent is sufficient for 10 blots.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service