Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5743

Sigma-Aldrich

Anti-PC-Specific PLD1, N-Terminal antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Phosphatidylcholine Specific Phospholipase-D1, N-Terminal

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

~0.5 mg/mL

technique(s)

immunoprecipitation (IP): suitable
western blot: 1:1,000 using recombinant purified PLD1 protein

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... PLD1(5337)

Specificity

Detects human recombinant and natural PLD1.

Immunogen

synthetic peptide derived from the N-terminal region of human PC-specific PLD1 protein (aa 1-15).

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 0.1% BSA and 0.05% sodium azide.

Analysis Note

The amount of the reagent is sufficient for 10 blots.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William E Hughes et al.
The Journal of biological chemistry, 279(26), 27534-27541 (2004-04-17)
Phospholipase D (PLD) has been strongly implicated in the regulation of Golgi trafficking as well as endocytosis and exocytosis. Our aim was to investigate the role of PLD in regulating the biphasic exocytosis of insulin from pancreatic beta-cells that is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service