Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5623

Sigma-Aldrich

Anti-Potassium Channel KIR3.4 (GIRK4) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-G Protein-activated Inward Rectifier K+ Channel 4, Anti-GIRK4, Anti-KCNJ5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, human, rat

technique(s)

western blot: 1:200 using rat heart membranes

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ5(3762)
mouse ... Kcnj5(16521)
rat ... Kcnj5(29713)

Immunogen

peptide, RNAMNQDMEIGVT(C), corresponding to residues 6-18 of rat Kir3.4 (GIRK4). The sequence is identical in rat, human, pig, and mouse.

Physical form

Lyophilized powder from phosphate buffered saline containing 1% bovine serum albumin, and 0.025% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service