Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5578

Sigma-Aldrich

L-α-Phosphatidyl-D-myo-inositol 3-monophosphate, dioctanoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H48O16P2
Molecular Weight:
666.59
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

CCCCCCCC(=O)OC[C@H](COP(O)(=O)O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](OP(O)(O)=O)[C@H]1O)OC(=O)CCCCCCC

InChI

1S/C25H48O16P2/c1-3-5-7-9-11-13-18(26)37-15-17(39-19(27)14-12-10-8-6-4-2)16-38-43(35,36)41-25-22(30)20(28)21(29)24(23(25)31)40-42(32,33)34/h17,20-25,28-31H,3-16H2,1-2H3,(H,35,36)(H2,32,33,34)/t17-,20+,21+,22-,23-,24-,25+/m1/s1

InChI key

LKXJHTKDXMQMDM-UCGOVMIJSA-N

Substrates

Component of lipid signaling pathway; formed from phosphatidylinositol by phosphatidylinositol 3-kinase.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Huang et al.
Nature, 391(6669), 803-806 (1998-03-05)
Inward rectifier K+ channels, which modulate electrical activity in many cell types, are regulated by protein kinases, guanine-nucleotide-binding proteins (G proteins) and probably actin cytoskeleton. Generation of phosphatidylinositol 4,5-bisphosphate (PIP2) by ATP-dependent lipid kinases is known to activate inward rectifier
J A Pitcher et al.
The Journal of biological chemistry, 270(20), 11707-11710 (1995-05-19)
The pleckstrin homology (PH) domain is an approximately 100-amino-acid region of sequence homology present in numerous proteins of diverse functions, which forms a discrete structural module. Several ligands capable of binding to PH domain-containing proteins have been identified including phosphatidylinositol
J E Harlan et al.
Biochemistry, 34(31), 9859-9864 (1995-08-08)
The pleckstrin homology (PH) domain is a protein module of approximately 100 amino acids that is found in several proteins involved in signal transduction [for a recent review, see Gibson et al. (1994) Trends Biochem. Sci. 19, 349-353]. Although the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service