Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5297

Sigma-Aldrich

Palmitoyl coenzyme A–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

saline suspension

extent of labeling

1-3 mg per mL

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

7 atoms

storage temp.

2-8°C

Application

Palmitoyl coenzyme A-agarose is used in protein chromatography and affinity chromatography. Palmitoyl coenzyme A-agarose has been used to identify the human mitochondrial linoleoyl-coenzyme A monolysocardiolipin acyltransferase (MLCL AT-1).

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tong Guo et al.
The Journal of cell biology, 177(2), 289-303 (2007-04-18)
We define the dynamics of spatial and temporal reorganization of the team of proteins and lipids serving peroxisome division. The peroxisome becomes competent for division only after it acquires the complete set of matrix proteins involved in lipid metabolism. Overloading
F Domergue et al.
Lipids, 35(5), 487-494 (2000-07-25)
Oleoyl-CoA elongase catalyzes four successive reactions: condensation of malonyl-CoA to oleoyl-CoA, reduction, dehydration, and another reduction. Evidence supporting this mechanism and the multienzymatic nature of the elongation complex are reported. A particulate membrane fraction from rapeseed is able to elongate
Abel Hiol et al.
Biochimica et biophysica acta, 1635(1), 10-19 (2003-12-04)
The acylation of proteins through the addition of palmitate to cysteine residues is a common posttranslational modification for a variety of proteins, but the enzymology of this reversible modification has resisted elucidation. We developed a strategy to purify protein fatty
William J Driscoll et al.
The Journal of biological chemistry, 282(31), 22353-22363 (2007-05-15)
Oleamide (cis-9-octadecenamide) is the prototype member of an emerging class of lipid signaling molecules collectively known as the primary fatty acid amides. Current evidence suggests that oleamide participates in the biochemical mechanisms underlying the drive to sleep, thermoregulation, and antinociception.
M Elholm et al.
The Journal of biological chemistry, 276(24), 21410-21416 (2001-03-30)
The peroxisome proliferator-activated receptor alpha (PPARalpha) is a ligand-activated transcription factor and a key regulator of lipid homeostasis. Numerous fatty acids and eicosanoids serve as ligands and activators for PPARalpha. Here we demonstrate that S-hexadecyl-CoA, a nonhydrolyzable palmitoyl-CoA analog, antagonizes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service