P5233
Hamilton® Softgrip® adjustable volume pipette
volume 10-100 μL, resolution: 0.2 μL
About This Item
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Hamilton 55019-36
10% volume accuracy measuring range
accuracy: 1.80%, precision: 0.70%
100% volume accuracy measuring range
accuracy: 0.60%, precision: 0.15%
volume measuring range
10-100 μL, resolution: 0.2 μL
volume
100 μL
color
violet
General description
The separate, easy-to-reach, tip ejector button is yet another convenient feature of these state-of-the-art pipettes. You′ll feel these proven differences:
- 50% less actuation force with a corresponding reduction in risk factors associated with musculoskeletal disorders, such as carpal tunnel syndrome
- Less user fatigue due to the soft ergonomic shape and lower plunger forces
- Less discomfort in the hand and wrist as compared to other pipettes during prolonged use
Every SoftGrip pipette is calibrated at Hamilton with standards traceable to NIST. A certificate of calibration documenting accuracy and precision is provided in the package along with non-wettable recalibration labels for field service and recalibration.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)