Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4996

Sigma-Aldrich

Anti-Potassium Channel EAG-2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Ether-a-go-go Potassium Channel 2, Anti-KV10.2, Anti-KCNH5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... KCNH5(27133)
rat ... Kcnh5(171146)

Immunogen

peptide corresponding to amino acid residues 842-860 of rat EAG-2. This epitope is highly homologous in human (18/19 residues identical).

Physical form

Lyophilized at ~0.8 mg/mL in phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.05 % sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service