Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4995

Sigma-Aldrich

Anti-PRMT4 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CARM1, Anti-Protein-Arginine Methyl Transferase 4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 5-20 μg using PRMT4-fusion protein from extracts of transfected 293T cells.
microarray: suitable
western blot: 0.5-1 μg/mL using nuclear extracts of the MCF7 breast carcinoma cell line.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CARM1(10498)
mouse ... Carm1(59035)

General description

Protein arginine methyltransferase 4 (PRMT4), which is also known as coactivator associated arginine methyltransferase 1 (CARM1), is part of the protein arginine methyltransferase (PRMT) family. The gene encoding this protein is localized on human chromosome 19p13.2.

Immunogen

synthetic peptide corresponding to amino acids 48-63 of human PRMT4 conjugated to KLH via a C-terminal added lysine residue. The sequence is conserved in mouse.

Application

Anti-PRMT4 antibody produced in rabbit has been used for Western Blotting and immunofluorescence.

Biochem/physiol Actions

Protein arginine methyltransferase 4 (PRMT4) methylates arginine residues of histones and other proteins and thus has a role in modulation of gene expression. The protein also methylates histone acetyl transferases. It has been shown to associate with Mi2α, a chromatin remodeler. It also associates with nuclear factor κB (NF-κB), estrogen receptor and tumor suppressor p53 and is recruited to specific target genes. PRMT4 has a role in pre-mRNA splicing and DNA damage response. The protein has been shown to be upregulated in liver, prostate and breast cancers.
Protein-arginine methyl transferase 4 (PRMT4) plays a central role in upregulation of transcription by nuclear hormone receptors (NRs).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coactivator-Associated Arginine Methyltransferase 1 Enhances Transcriptional Activity of the Human T-Cell Lymphotropic Virus Type 1 Long Terminal Repeat through Direct Interaction with Tax
Jeong SJ, et al.
Journal of Virology, 80, 10036-10044 (2006)
High-resolution genomic profiling of Philadelphia chromosome-positive (Ph+) acute lymphoblastic leukemia (ALL) patients identified novel recurrent copy number variations involved in both pathogenesis and resistance to tyrosine kinase inhibitors
Ilaria Iacobucci
Cancer Research, 68(9) (2008)
Mapping of Post-translational Modifications of Transition Proteins, TP1 and TP2, and Identification of Protein Arginine Methyltransferase 4 and Lysine Methyltransferase 7 as Methyltransferase for TP2*
Nikhil Gupta
The Journal of Biological Chemistry, 12101-12122 (2015)
CARM1 Preferentially Methylates H3R17 over H3R26 through a Random Kinetic Mechanism.
Jacques SL
Biochemistry, 55(11), 1635-1644 (2016)
Nikhil Gupta et al.
The Journal of biological chemistry, 290(19), 12101-12122 (2015-03-31)
In a unique global chromatin remodeling process during mammalian spermiogenesis, 90% of the nucleosomal histones are replaced by testis-specific transition proteins, TP1, TP2, and TP4. These proteins are further substituted by sperm-specific protamines, P1 and P2, to form a highly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service