P4868
Anti-p53DINP1/SIP antibody produced in rabbit
~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-Stress Induced Protein, Anti-p53-Dependent Damage-Inducible Nuclear Protein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352203
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
rat, human, mouse
concentration
~1 mg/mL
technique(s)
microarray: suitable
western blot: 1-2 μg/mL
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... TP53INP1(94241)
mouse ... Trp53inp1(60599)
rat ... Tp53inp1(297822)
Specificity
Recognizes a band at 27 kDa. In human spleen, mouse liver, and mouse kidney tissue lysates, a band at 18 kDa may be detected.
Immunogen
synthetic peptide corresponding to amino acids 1-14 of human p53DINP1.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in phosphate buffered saline containing 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rebeca Rosas-Hernández et al.
Journal of neurochemistry, 149(2), 242-254 (2018-12-28)
Alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid glutamate receptors have been shown to modulate the morphology of the lamelar processes of Bergmann glia cells in the molecular layer of the cerebellum. Here we suggest that reorganization of F-actin may underlay the changes in the morphology
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service