Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4746

Sigma-Aldrich

Anti-Purinergic Receptor P2X6 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-P2rxl1, Anti-P2x6, Anti-Rnap2x6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide corresponding to amino acid residues 363-379 of rat P2X6. This epitope is highly homologous in mouse (15/17 residues identical) and human (14/17 residues identical).

Physical form

Lyophilized in phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service