Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4661

Sigma-Aldrich

pH test strips

pH 0.0-6.0, resolution: 0.5 pH unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.79

feature

non-bleeding: no
refill: no

pH measuring range

0.0-6.0, resolution: 0.5 pH unit

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

General description

Non-bleeding pH color strips. Simply immerse the strip in the test solution until color development is complete. Then compare the developed color to the sequence chart on the package, and read the pH. The range covered by each type of strip is divided into graduations as given below. Each package contains 100 strips in a clear, plastic container with a snug fitting top.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service