P4656
Pepsinogen from porcine stomach
Grade I-S, lyophilized powder, ~3,000 units/mg protein (after activation to pepsin at pH 2.0 at 25°C)
Synonym(s):
Pepsinogen from hog stomach
About This Item
Recommended Products
biological source
Porcine stomach
type
Grade I-S
assay
≥85% (biuret)
form
lyophilized powder
specific activity
~3,000 units/mg protein (after activation to pepsin at pH 2.0 at 25°C)
purified by
chromatography
UniProt accession no.
foreign activity
pepsin, free
storage temp.
2-8°C
Gene Information
pig ... PGA5(396892)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Unit Definition
Analysis Note
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service