P4631
Phosphodiesterase I from Bothrops atrox
Type V, crude dried venom, ≥0.01 unit/mg solid
Synonym(s):
5′-Exonuclease, Oligonucleate 5′-nucleotidohydrolase
About This Item
Recommended Products
type
Type V
form
crude dried venom
specific activity
≥0.01 unit/mg solid
storage temp.
−20°C
Unit Definition
inhibitor
substrate
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service