Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4622

Sigma-Aldrich

Anti-Potassium Channel Kv3.3 (KNC3) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, human, mouse

technique(s)

western blot: 1:200 using rat brain membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... KCNC3(3748)
mouse ... Kcnc3(16504)
rat ... Kcnc3(117101)

Specificity

Recognizes splice variant KV3.3-1.

Immunogen

synthetic peptide corresponding to amino acid residues 701-718 of rat KV3.3. The sequence is identical in human and mouse.

Physical form

Lyophilized powder from phosphate buffered saline containing 1% bovine serum albumin and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service