Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4496

Sigma-Aldrich

Anti-Potassium Channel Kir2.2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-IRK2, Anti-Kcnj12

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
200 μL
$1,040.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot: 1:200 using rat kidney and brain membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ12(3768)
mouse ... Kcnj12(16515)
rat ... Kcnj12(117052)

Immunogen

peptide corresponding to amino acid residues 391-409 of rat Kir2.2. This epitope is highly homologous in mouse (16/19 residues identical) and human (13/19 residues identical).

Physical form

Lyophilized at ~0.5 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service