Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4488

Sigma-Aldrich

Pancreatic Polypeptide Fragment 1-17–[Ala31, α-Aminoisobutyryl32]-Neuropeptide Y Fragment 18-36

≥95% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C189H287N55O53S
Molecular Weight:
4209.70
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... NPY(4852)

Amino Acid Sequence

Ala-Pro-Leu-Glu-Pro-Val-Tyr-Pro-Gly-Asp-Asn-Ala-Thr-Pro-Glu-Gln-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ala-Aib-Arg-Gln-Arg-Tyr-NH2

Biochem/physiol Actions

Selective agonist of the neuropeptide Y receptor.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service