Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4385

Sigma-Aldrich

Calmodulin−Agarose

saline suspension

Synonym(s):

Phosphodiesterase 3′,5′-Cyclic Nucleotide Activator-Agarose from bovine testes

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500

form

saline suspension

extent of labeling

≥0.5 mg per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Calmodulin-agarose is used in affinity chromatography, protein chromatography, calcium associated proteins, and specialty resins. Calmodulin-agarose has been used to suggest a coordinated regulation of processes in both mitochondria and chloroplasts by calcium signals. Calmodulin-agarose has also been used to study apoptosis in prostate cancer cells and to analyze the interaction of the influenza virus polymerase complex with human cell factors.

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Takasaki et al.
The Journal of biological chemistry, 274(17), 11848-11853 (1999-04-17)
Various proteins in the signal transduction pathways as well as those of viral origin have been shown to be myristoylated. Although the modification is often essential for the proper functioning of the modified protein, the mechanism by which the modification
M S Rogers et al.
Neoplasia (New York, N.Y.), 1(3), 220-225 (2000-08-10)
Cell proliferation requires calmodulin, a protein that regulates calcium-dependent enzymes involved in signal transduction pathways in eukaryotic cells. Calmodulin-like protein (CLP) is found in certain epithelial cell types, including normal breast epithelium, and, although it closely resembles calmodulin in amino
L Luoni et al.
FEBS letters, 482(3), 225-230 (2000-10-12)
The plasma membrane Ca(2+)-ATPase was purified from Arabidopsis thaliana cultured cells by calmodulin (CaM)-affinity chromatography and reconstituted in proteoliposomes by the freeze-thaw sonication procedure. The reconstituted enzyme catalyzed CaM-stimulated 45Ca(2+) accumulation and H(+) ejection, monitored by the increase of fluorescence
Daisuke Takezawa et al.
Biochemical and biophysical research communications, 317(2), 428-436 (2004-04-06)
Plant responses to environmental stresses are mediated in part by signaling processes involving cytosolic Ca2+ and a Ca(2+)-binding protein, calmodulin. Screening with radiolabeled calmodulin of a cDNA library of the moss Physcomitrella patens resulted in identification of genes encoding novel
Daisuke Takezawa
The Journal of biological chemistry, 278(39), 38076-38083 (2003-07-16)
Protein phosphatases regulated by calmodulin (CaM) mediate the action of intracellular Ca2+ and modulate functions of various target proteins by dephosphorylation. In plants, however, the role of Ca2+ in the regulation of protein dephosphorylation is not well understood due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service