P4366
Nunc® GeNunc™ PCR Amplifcation Workstation
autoclavable (plates only), w/lid
Synonym(s):
Nunc PCR workstation, Nunc amplification workstation, Nunc tube tray and holder, PCR amplification workstation, PCR tube rack, PCR workstation
About This Item
feature
autoclavable (plates only)
lid (not autoclavable)
packaging
pack of 1
case of 5
manufacturer/tradename
Nunc 251357
technique(s)
PCR: suitable
General description
- Workstation for PCR set-up
- Easy to assemble
- Standard 96 MicroWell format to hold 0.2 mL PCR tubes, strips or Nunc 96 Well Amplifications
- Plate compatible with automated handling systems
- Removable tray can be fitted directly into the thermal cycler
- Compatible with V-bottom 0.2 mL tube block thermal cycler formats of major manufacturers
- Alphanumerically marked for sample identification
- Tray fits holder in only one way to make orientation easy
- Can be used as a storage system
- Stackable space saving units with lid lugs for stability during storage
- Can be used from -20°C to +110°C
- Chemically resistant to weak acids and alcohols
The following thermocylcers are compatible with the GeNunc PCR Amplification Workstation:
Nunc′s GeNunc amplification workstation is compatible with 96 well format thermocyclers with a 0.2 mL tube block. For example, Perkin Elmer Gene Amp PCR System 9600, Perkin Elmer Gene Amp PCR System 9700, Techne Genius with 96 x 0.2 mL block, MJ Research PTC-200 DNA Engine, MJ Research PTC-100-96, Ericomp Delta Cycler, Barnstead Thermolyne Amplitron II, Lab-Line Programmable Thermal Blok Model 212 and Hybaid Touchdown with 0.2 mL block are each compatible with the workstation.
The GeNunc PCR Amplification Workstation is made of the following material:
Polyphenylenine oxide. This material is resistant to solvents and weak acids.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service