Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4182

Sigma-Aldrich

N,N′,N′-Polyoxyethylene (10)-N-tallow-1,3-diaminopropane

liquid

Synonym(s):

EDT-20, PEG-10 tallow aminopropylamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900

description

cationic

form

liquid

color

dark amber

General description

N,N′,N′-Polyoxyethylene (10)-N-tallow-1,3-diaminopropane (EDTA-20) is a cationic detergent. [1]

Application

N,N′,N′-Polyoxyethylene (10)-N-tallow-1,3-diaminopropane has been used in a study to demonstrate that PC2 facilitates the activation of transcription by GAL4-AH. [2] It has also been used in a study to investigate optimum in vitro excystation of Notocotylus attenuatus. [3] It is also useful for extraction of enzymes from microbial cells.

Quantity

Recommended usage: 1 ml per 100 grams wet cell weight buffered at pH 7.3-8.0.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kretzschmar et al.
Molecular and cellular biology, 14(6), 3927-3937 (1994-06-01)
We have isolated from a crude Hela cell cofactor fraction (USA) a novel positive cofactor that cooperates with the general transcription machinery to effect efficient stimulation of transcription by GAL4-AH, a derivative of the Saccharomyces cerevisiae regulatory factor GAL4. PC2
Excystment in vitro of Notocotylus attenuatus (Trematoda: Notocotyloidea) metacercariae
Thaddeus K Graczyk and Clive J Shiff
J. Parisitol., 79, 92-92 (1993)
Extraction of Haemoproteus columbae (Haemosporina: Haemoproteidae) antigen from rock dove pigeons (Columba livia) and its use in an antibody ELISA
Thaddeus K. Graczyk et al.
J. Parisitol., 80, 713-713 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service