P3987
Anti-Protein Kinase D antibody produced in rabbit
IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-PKCμ, Anti-PKD
About This Item
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 110 kDa
species reactivity
human, mouse
technique(s)
immunoprecipitation (IP): 10 μL using cell lysate of mouse fibroblast NIH3T3 cell line.
microarray: suitable
western blot: 1:500 using whole cell extract of human epidermal carcinoma A431 cell line.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... PRKD1(5587)
General description
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service