Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3899

Sigma-Aldrich

Poly(Glu, Ala, Tyr) sodium salt

Glu:Ala:Tyr (6:3:1), mol wt 20,000-50,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

form

powder or solid

feed ratio

Glu:Ala:Tyr (6:3:1)

mol wt

20,000-50,000

technique(s)

bioconjugation: suitable

color

white to off-white

application(s)

cell analysis

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Seo et al.
The Journal of investigative dermatology, 116(6), 880-885 (2001-06-16)
The physiologic role of gamma delta-T-cell-receptor-bearing cells and the T cell receptor ligands that they recognize is still poorly understood. Previous studies have suggested that one possible antigen for gamma delta-TCR(+) cells is the random copolymer poly-glutamic acid-tyrosine (poly-Glu-Tyr), because

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service