Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3735

Sigma-Aldrich

Anti-p62dok antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 62 kDa

species reactivity

human

concentration

~0.5 mg/mL

technique(s)

western blot: 1:1,000-1:2,000 using whole cell lysate of human Jurkat cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... DOK1(1796)

Immunogen

synthetic peptide corresponding to amino acids 425-439 of human p62dok.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service