Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3616

Sigma-Aldrich

[pTyr783]-PLCγ

≥80% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

Product Name

[pTyr783]-PLCγ, ≥80% (HPLC), solid

assay

≥80% (HPLC)

form

solid

mol wt

1790

composition

Peptide content, ≥75%

solubility

H2O: soluble

storage temp.

−20°C

Biochem/physiol Actions

Positive control peptide for the [Tyr783]-PLCγ rabbit polyclonal antibody.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antiangiogenic-Induced Hypertension: The Molecular Basis of Signaling Network.
Nagai A, Sado T, Naruse K, et al.
Gynecologic and Obstetric Investigation (2012)
Natsuko Kageyama-Yahara et al.
Biochemical and biophysical research communications, 416(3-4), 283-288 (2011-11-22)
Antigen-IgE-mediated mucosal mast-cell activation is critical in the development of food allergies. Cinnamaldehyde, a major constituent of Cinnamomi cortex, dose-dependently inhibited the antigen-IgE-induced degranulation of mucosal-type bone-marrow derived mast cells (mBMMCs) and RBL-2H3 cells. Cinnamaldehyde also suppressed the elevation of
J W Li et al.
Journal of genetics, 90(3), 435-442 (2012-01-10)
Although it is known that hormones, growth factors and integrin promote hepatocyte proliferation in liver regeneration (LR) through ERK1/2 signalling pathway, reports about regulating processes of its intracellular paths in hepatocytes of LR are limited. This study aims at exploring
Hee Jun Cho et al.
Biochemical and biophysical research communications, 414(3), 575-580 (2011-10-12)
Rho GDP dissociation inhibitor 2 (RhoGDI2) is a regulator of the Rho family GTPases. Recent work from our laboratory suggests that RhoGDI2 expression potentially enhances resistance to cisplatin as well as promotes tumor growth and malignant progression in gastric cancer.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service