Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3319

Sigma-Aldrich

Penicillin Amidase from Escherichia coli

ammonium sulfate suspension, ≥10 units/mg protein (E1%/280)

Synonym(s):

Penicillin Acylase, Penicillin Amidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

biological source

Escherichia coli

Quality Level

form

ammonium sulfate suspension

specific activity

≥10 units/mg protein (E1%/280)

shipped in

wet ice

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will hydrolyze 1.0 μmole of benzylpenicillin per min at pH 7.8 at 37°C.

Physical form

Suspension in 0.1 M phosphate, pH 7.5 and 3 M ammonium sulfate

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronese, F., et al.
Il Farmaco Ed. Sc., 36, 663-663 (1981)
Nishant Kumar Varshney et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 68(Pt 3), 273-277 (2012-03-24)
The enzyme penicillin G acylase (EC 3.5.1.11) catalyzes amide-bond cleavage in benzylpenicillin (penicillin G) to yield 6-aminopenicillanic acid, an intermediate chemical used in the production of semisynthetic penicillins. A thermostable penicillin G acylase from Alcaligenes faecalis (AfPGA) has been crystallized
Özlem Akkaya et al.
World journal of microbiology & biotechnology, 28(5), 2159-2164 (2012-07-19)
Penicillin G acylase (PA) is an important enzyme used in the industrial production of b-lactam antibiotics. In this study, the effects of mutations in the translation initiation region of the Escherichia coli pac gene, encoding periplasmic PA, were examined. Several
Pedro Valencia et al.
New biotechnology, 29(2), 218-226 (2011-09-20)
A mathematical model is presented for the kinetically controlled synthesis of cephalexin that describes the heterogeneous reaction-diffusion process involved in a batch reactor with glyoxyl-agarose immobilized penicillin acylase. The model is based on equations considering reaction and diffusion components. Reaction
Jun Yin et al.
The Journal of biological chemistry, 286(27), 24476-24486 (2011-05-18)
Cephalosporin acylase (CA) precursor is translated as a single polypeptide chain and folds into a self-activating pre-protein. Activation requires two peptide bond cleavages that excise an internal spacer to form the mature αβ heterodimer. Using Q-TOF LC-MS, we located the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service