Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3203

Sigma-Aldrich

Anti-Protein Kinase Cβ2 antibody produced in rabbit

0.5 mg/mL, affinity isolated antibody

Synonym(s):

Anti-PKC β2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen 80 kDa

species reactivity

rat

concentration

0.5 mg/mL

technique(s)

dot blot: 1:1,000
microarray: suitable
western blot: 1:8,000 using rat brain extract

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Protein Kinase C (PKC, 76-93 kD) is a family of Ser/Thr specific protein kinases that perform key functions in numerous signalling pathways, in biological systems, through their various isoforms. The conventional PKC isoforms (cPKC) are PKC-α, β1, β2 and γ; activated by phosphatidylserine, calcium or phorbol esters. Proteolysis of PKC in vivo is thought to be mediated by calpains I and II. Calpains cleave PKC in the V3 hinge region to produce two distinct fragments, one comprising the N-terminal regulatory domain (30 kD) and the other fragment containing the C-terminal kinase domain (50 kD) that is catalytically active. Multiple functions such as, cellular and vascular regulations, angiogenesis, cell growth, apoptosis, changes in basement membrane thickness, extracellular matrix organisation, MAPK signalling, are attributed to PKC isoforms. These varied functions implicate PKC isoforms in cardiac hypertrophies and diabetic nephropathy and cardiovascular complications.
Anti-Protein Kinase C β2 antibody specifically recognises PKC β2 (80 kDa).

Specificity

In immunoblotting, the product reacts with PKC β2 (80 kDa) using rat brain extract.

Immunogen

synthetic peptide corresponding to the C-terminal region of PKC β2 (660-673).

Application

Anti-Protein Kinase C β2 antibody may be used for detection by immunoblotting at a working dilution of 1:8000 using rat brain extract. It may be used in dat blot at a working dilution of 1:1000. It is suitable for protein microarray.

Physical form

Supplied in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Preparation Note

Purified by affinity chromatography using immobilized PKC β2 peptide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Abu-Ghanem et al.
Cancer biology & therapy, 6(9), 1375-1380 (2007-09-06)
The Hodgkin-Reed-Sternberg (HRS) malignant cells in Hodgkin's lymphoma (HL) originate from germinal center B lymphocytes that did not undergo apoptosis. Protein Kinase C (PKC), a family of serine/threonine kinases, plays a crucial role in signal transduction modulating cell growth, differentiation
Weiqin Yin et al.
Journal of diabetes investigation, 10(3), 613-625 (2018-10-12)
Blockade or reversal the progression of diabetic nephropathy is a clinical challenge. The aim of the present study was to examine whether recombinant human glucagon-like peptide-1 (rhGLP-1) has an effect on alleviating urinary protein and urinary albumin levels in diabetic
Domain interactions in protein kinase C.
C J Pears et al.
Journal of cell science, 100 ( Pt 4), 683-686 (1991-12-01)
S Budhiraja et al.
Fundamental & clinical pharmacology, 22(3), 231-240 (2008-05-20)
Diabetic nephropathy (DN) has emerged as the major causative pathology in patients entering end-stage renal disease (ESRD) worldwide and it is responsible for 30-40% of all ESRD cases. Treatments for DN are centered on control of hyperglycemia and blood pressure
Alexandra C Newton
The Biochemical journal, 370(Pt 2), 361-371 (2002-12-24)
Phosphorylation plays a central role in regulating the activation and signalling lifetime of protein kinases A, B (also known as Akt) and C. These kinases share three conserved phosphorylation motifs: the activation loop segment, the turn motif and the hydrophobic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service