Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3104

Sigma-Aldrich

p13suc1-Agarose

extent of labeling: 2.5 mg GST-p13suc1 per 0.5 mL resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

extent of labeling

2.5 mg GST-p13suc1 per 0.5 mL resin

shipped in

wet ice

storage temp.

2-8°C

Specificity

May be used for isolation of p34cdc2.

Physical form

Suspension in phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Preparation Note

Prepared using recombinant GST-p13suc1 (approximately 39 kDa) non-covalently bound to glutathione-agarose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service