Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3098

Sigma-Aldrich

(−28) M13 Reverse primer set

Synonym(s):

5′-AGG AAA CAG CTA TGA CCA T-3′

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305

storage temp.

2-8°C

Application

(−28) M13 Reverse primer set is a single-stranded oligonucleotide with 5′-hydroxyl and 3′-hydroxyl ends and a selection of four fluorescent lables for use in polymerase chain reaction protocols (PCR).

Packaging

Packaged in amber tubes to protect from light.

Components

Set of four vials of primer, each individually labeled at the 5′-end with:
FAM: (5[6]-carboxyfluorescein)
JOE: (6-carboxy-4′,5′-dichloro-2′,7′-dimethoxyfluorescein)
ROX: (5[6]-carboxy-X-rhodamine), and
TAMRA: (5[6]-carboxytetramethylrhodamine.
Supplied as solutions, 50 μl each, in tris-EDTA buffer, pH 7.5, at 1 picomole/μl

Analysis Note

Tested for purity with HPLC, PAGE, and OD.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K A Abd-Elsalam et al.
Genetics and molecular research : GMR, 9(4), 2016-2024 (2010-10-20)
Seven fungal isolates were identified as pan-global Hypocrea/Trichoderma species, from section Trichoderma, on the basis of their morphology. These species were H. lixii/T. harzianum and H. orientalis/T. longibrachiatum. PCR-based markers with primer M13 (core sequence of phage M13) and internal-transcribed
Angela Capece et al.
International journal of food microbiology, 144(1), 187-192 (2010-10-12)
The present research studied Saccharomyces cerevisiae yeasts isolated from Nero d'Avola grapes, collected in different areas of the Sicily region. RAPD-PCR analysis with M13 primer was used for preliminary discrimination among 341 S. cerevisiae isolates. Inoculated fermentations with S. cerevisiae
High-throughput sequencing of PCR products tagged with universal primers using 454 life sciences systems.
Daigle D, Simen BB, Pochart P.
Current Protocols in Molecular Biology, Unit 7-Unit 7 (2011)
Yajing Zhou et al.
PloS one, 6(7), e22224-e22224 (2011-07-27)
Eukaryotic DNA polymerase δ (pol δ) plays a crucial role in chromosomal DNA replication and various DNA repair processes. It is thought to consist of p125, p66 (p68), p50 and p12 subunits. However, rigorous isolation of mammalian pol δ from
Jacquelina Williams-Woods et al.
Journal of virological methods, 178(1-2), 253-257 (2011-10-04)
Human norovirus (HuNoV) and hepatitis A (HAV) are recognized as leading causes of non-bacterial foodborne associated illnesses in the United States. DNA sequencing is generally considered the standard for accurate viral genotyping in support of epidemiological investigations. Due to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service